Learn More
Aviva Systems Biology Corp HOXA2 Antibody - middle region (ARP31950_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31950_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HOXA2. Alternate names: HOX1KApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52846