Learn More
Medchemexpress LLC HP211206 | 3119106-17-8 | 883.94 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1705981MG
Encompass_Preferred
HP211206 is a SARS-CoV-2 main protease (Mpro) PROTAC degrader. It effectively degrades SARS-CoV-2 Mpro and its drug-resistant mutants. It exhibits an IC50 of 181.9 nM and a DC50 of 621 nM for Mpro, and it also possesses antiviral activity. The compound is composed of H117 (HY-170599) in pink, a linker (HY-B0236) in black, and an E3 ligase ligand (HY-41547) in blue.
  • SARS-CoV-2 Mpro PROTAC degrader.
  • Effectively degrades SARS-CoV-2 Mpro and its drug-resistant mutants.
  • Has an IC50 of 181.9 nM for Mpro.
  • Has a DC50 of 621 nM for Mpro.
  • Exhibits antiviral activity.
Catalog No. 50-003-85734