Learn More
Cayman Chemical HPI-1hydrate 50mg
SDP
Supplier:  Cayman Chemical 1522150 MG
Encompass_Preferred
A Hh pathway inhibitor that suppresses signaling through Sonic Hh (IC50 = 1.5 µM) without affecting Wnt signaling (IC50 ≥ 30 µM); suppresses Hh activation induced by loss of Suppressor of Fused or by Gli overexpression; also inhibits signaling through the oncogenic mutant SmoM2
Catalog No. 50-297-0736