Learn More
Medchemexpress LLC Hpk1-in-2 dihydrochloride | 2375595-72-3 | 98.7% | 453.39 g·mol⁻¹ | C19H22Cl2N6OS | 5 MG

Supplier: Medchemexpress LLC HY132150A5MG
HPK1-IN-2 dihydrochloride is a small-molecule research inhibitor of hematopoietic progenitor kinase 1 (HPK1), supplied as the dihydrochloride salt for use in in vitro and preclinical studies. It is used to probe T-cell signaling and evaluate antitumor activity, and is supplied as a high-purity solid or ready-to-use solution for assay applications.
- Potent HPK1 inhibitor for in vitro and preclinical research
- CAS number 2375595-72-3 for unambiguous identification
- Molecular formula C19H22Cl2N6OS; molecular weight 453.39 g·mol⁻¹
- Recommended purity 98.69% (typical)
- Appearance light brown to brown solid
- Storage: 4°C sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month)
- Available in small pack sizes and DMSO solution for convenient assay setup
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.