Learn More
Medchemexpress LLC Hpk1-in-34 | 2380300-99-0 | C25H28N4O2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15336210MM/1ML
Encompass_Preferred
HPK1-IN-34 is an inhibitor of Hematopoietic progenitor kinase 1 (HPK1) with an IC50 of less than 100 nM. It is also a click chemistry reagent, featuring an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups. This product is intended for research use only.
  • Inhibitor of HPK1 with an IC50 of less than 100 nM
  • Click chemistry reagent
  • Features an alkyne group for CuAAc reactions
  • Light yellow to yellow solid
  • Intended for research use only
Catalog No. 50-003-72866