Learn More
Microsolv Technology Corporation HPLC Column, Amide, 4um, 4.6mm ID x 50mm Length, 100A. 1 EA.
Supplier:  Microsolv Technology Corporation 4003605P
Encompass_Preferred
This Cogent Amide HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of Sugars and other Analytes with amine groups, This size column provides good resolution and short run times.
Catalog No. 50-227-0757