Learn More
Perkin Elmer US LLC HPLC Column CAPCELL PAK C18 MG 100A 5µm, 250 mm x 4.6 mm
Supplier:  Perkin Elmer US LLC N8145326
Encompass_Preferred
HPLC Column CAPCELL PAK C18 MG 100A Packing: C18 (RP18, ODS, Octadecyl) Pore Size: 100A Length range: 201 - 300
Catalog No. 50-209-3303