Learn More
Microsolv Technology Corporation HPLC Column, HPS Silica, 5um, 4.6mm ID x 100mm Length, 120A. 1 EA.
Supplier:  Microsolv Technology Corporation 7500110P
Encompass_Preferred
This Cogent HPS Silica™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. This proprietary Silica is not modified in any way and is ideal as an Alternate in any USP Method that uses the L3 Classification or for new methods. With a 5 µm Particle, this column works well with higher flow rates and lower back pressures compared with smaller particles and is often used for hydrophilic or polar compounds in Normal Phase HPLC. Compare its performance to Inertsil and Luna products.
Catalog No. 50-227-1008