Learn More
Mac-Mod Avantor Partisil SAX, 85 A, 10 um, 4.6 x 250 mm HPLC Column (WVS hardware)
SDP
Supplier:  Mac-Mod 46821505
Encompass
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-304-793