Learn More
Mac-Mod Avantor Partisphere Silica, 120 A, 5 um, 4.6 x 125 mm HPLC Column (WVS Hardware)
SDP
Supplier:  Mac-Mod 46210501
Encompass
Avantor Partisphere is a high-purity gel packed in HPLC Columns. Avantor Partisphere columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-304-774