Learn More
Microsolv Technology Corporation HPLC Column, RP C18, 10um, 4.6mm ID x 50mm Length, 100A. 1 EA.
Supplier:  Microsolv Technology Corporation 6801805P
Encompass_Preferred
This Cogent RP C18™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available, making this column ideal as an Alternate in USP Methods that use the L1 Classification.With a 10 µm Particle, this column works well with higher flow rates and lower back pressures. Used for hydrophobic or neutral compounds in Reversed Phase, this Octadecyl Phase is excellent for long chain fatty acids. Expect longer retention than with a C8 column. This column is often chosen as a scouting column for preparative methods.
Catalog No. 50-226-9796