Learn More
Microsolv Technology Corporation HPLC Column, Silica-C 2.o, 2.2um, 2.1mm ID x 50mm Length, 120A, High Pressure. 1 EA.
Supplier:  Microsolv Technology Corporation 4020005P2
Encompass_Preferred
This Cogent Silica-C™ “near UHPLC”, HPLC Column is made with 2.2um, 120A, Cogent TYPE-C™ Silica , and this silica hydride based stationary phase. Chosen for analyzing compounds that are not soluble in Water including but not limited to: lipids, peptides, proteins, oligonucleotides and other compounds that require non polar solvents. Faster equilibration than other Silica columns, no need to control moisture in mobile phase solvents and robust column lifetimes This short and narrow bore column provides short run times but requires a pump that can precisely deliver, and a flow cell that made for, reduced flow rates with lower volumes.
Catalog No. 50-227-0818