Learn More
Microsolv Technology Corporation HPLC Column, Silica-C 2.o, 2.2um, 2.1mm ID x 150mm Length, 120A. 1 EA.
Supplier:  Microsolv Technology Corporation 4020015P2
Encompass_Preferred
This Cogent Silica-C 2.o™ “near UHPLC”, HPLC Column is made with 2.2um, 120A, Cogent TYPE-C™ Silica , and this silica hydride based stationary phase. Chosen for analyzing compounds that are not soluble in Water including but not limited to: lipids, peptides, proteins, oligonucleotides and other compounds that require non polar solvents. Faster equilibration than other Silica columns, no need to control moisture in mobile phase solvents and robust column lifetimes This column can be used for LCMS when increased resolution is desired.
Catalog No. 50-227-0820