Learn More
Microsolv Technology Corporation HPLC Column, UDA (wcx), 4um, 2.1mm ID x 100mm Length, 100A. 1 EA.
Supplier:  Microsolv Technology Corporation 4003110P2
Encompass_Preferred
This Cogent UDA™ (wcx) HPLC Column is made with Cogent TYPE-C™ Silica , and has an 11 carbon chain with a terminal carboxylic acid group. This silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Extremely Robust Column Lifetimes.Use in Reversed Phase (RP) , Aqueous Normal Phase (ANP) or as a Weak Cation Exchange Column (wcx at pH 6 or higher). The hydrophobic stationary phase has direct silicon-carbon bonds which makes the column very robust and can be used for hydrophilic or hydrophobic compounds, if desired. Also can be used at very low pH (below 2) without damage. This column is very popular and often chosen for LCMS when increased resolution is desired.
Catalog No. 50-227-0732