Learn More
Perkin Elmer US LLC HPLC Nut and Ferrule Kit - 0.0625 in, 2 ea.
Supplier:  Perkin Elmer US LLC A5530003
Encompass_Preferred
This kit includes the following pieces: 2 HPLC Ferrules, 1/16 inch (AX201594) 2 HPLC Bushings, 1/16 inch (AX201595)
Catalog No. 50-211-2274