Learn More
Trajan Scientific HPLC PROTECOL C18 Medical 3/PK
SDP
Supplier:  Trajan Scientific 2C18405T15K
Encompass_Preferred
C18G x 5 m particle size x 300 pore size 10 mm length x 150 m ID PEEKsil ProteCol HPLC trap column
Catalog No. 50-004-93700