Learn More
Trajan Scientific HPLC PROTECOL C8 Medical 3/PK
SDP
Supplier:  Trajan Scientific 2C8203T30K
Encompass_Preferred
C8H x 3 m particle size x 1000 pore size 10 mm length x 300 m ID PEEKsil ProteCol HPLC trap column
Catalog No. 50-004-94017