Learn More
Trajan Scientific HPLC PROTECOL Polar Medical 3/PK
SDP
Supplier:  Trajan Scientific 2POL05T30K
Encompass_Preferred
C18P x 5 m particle size x 120 pore size 10 mm length x 300 m ID PEEKsil ProteCol HPLC trap column
Catalog No. 50-004-94147