Learn More
Trajan Scientific HPLC Protecol UHPLC Column 1/EA
SDP
Supplier:  Trajan Scientific 08291131
Encompass
250 mm length x 2 1 mm ID ProteCol bio-inert UHPLC glass lined column 10-32 thread 0 5 m frit
Catalog No. 50-004-94234