Learn More
Trajan Scientific HPLC Protecol UHPLC Column 1/EA
SDP
Supplier:  Trajan Scientific 08291346
Encompass
100 mm length x 1 0 mm ID ProteCol bio-inert UHPLC glass lined column 6-32 thread 0 5 m frit
Catalog No. 50-004-94401