Learn More
Waters Corp Oasis Application Kits, HPLC, Therapeutic Peptide, Method Development
Supplier:  Waters Corp 176001836
Encompass_Preferred
The HPLC Therapeutic Peptide Method Development kit simplifies the process of sample preparation and LC method development for the analysis of therapeutic peptides in plasma. The kit contains a 96-well Oasis Peptide μElution Method Development Plate, an XBridge UPLC 3.5 μm BEH 300 C18 reversed-phase 2.1x50 mm column, three 96-well 1 mL collection plates and cap mats, and a detailed screening protocol.
  • Simplifies sample preparation and LC method development for therapeutic peptides in plasma.
  • Contains one 96-well peptide μElution method development plate.
  • Contains one UPLC 3.5 μm BEH 300 C18 reversed-phase 2.1x50 mm column.
  • Contains three 96-well 1 mL collection plates and cap mats.
  • Includes a detailed screening protocol.
Catalog No. 50-465-906