Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Supplier:  Trajan Scientific 2C18303N03P
Encompass_Preferred
C18Q x 3 m particle size x 100 pore size 150 mm length x 3 0 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-94009