Learn More
Trajan Scientific HPLC Reverse Phase Protecol C18 1/EA
SDP
Supplier:  Trajan Scientific 2C18505L02S
Encompass_Preferred
C18G x 5 m particle size x 120 pore size 50 mm length x 2 1 mm ID stainless steel ProteCol HPLC column
Catalog No. 50-004-94084