Learn More
Trajan Scientific HPLC Reverse Phase Protecol C8 1/EA
SDP
Supplier:  Trajan Scientific 2C8305R46P
Encompass_Preferred
C8H x 5 m particle size x 120 pore size 250 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-94141