Learn More
Trajan Scientific HPLC Reverse Phase Protecol Polar 1/EA
SDP
Supplier:  Trajan Scientific 2POL05R46P
Encompass_Preferred
C18P x 5 m particle size x 120 pore size 250 mm length x 4 6 mm ID PEEK coated ProteCol HPLC column
Catalog No. 50-004-93852