Learn More
Mac-Mod HydroBond AQ C8, 100 Å, 3 µm, 2.1 x 150 mm HPLC Column
SDP
Supplier:  Mac-Mod MMHB821153
Encompass
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-001-50948