Learn More
Mac-Mod Fortis BIO C8, 300 Å, 3 µm, .15 x 100 mm HPLC Column
SDP
Supplier:  Mac-Mod C15010003BIO308
Encompass
Fortis-BIO columns are manufactured with 300 A fully porous silica particles specifically designed for the retention and resolution of Peptide and Protein analyte types. C18 and C4 bonded phases are available on 1.7 and 5 um particle sizes for HPLC and UHPLC method development. Fortis BIO columns are available in a wide range of column IDs and column lengths including nano, capillary, microbore and analytical.
Catalog No. 50-001-52739