Learn More
Mac-Mod ProntoSIL Phenyl, 120 Å, 3 µm, 4.6 x 75 mm HPLC Column
SDP
Supplier:  Mac-Mod 0746F050PS030
Encompass
ProntoSIL HPLC columns produce great results, while at the same time being an economical column. Columns are packed in 400 bar HPLC hardware. There are several RPLC, HILIC, NPLC and Application Specific Columns with over 25 different bonded phases which are sure to solve your separation issues.
Catalog No. 50-001-48862