Learn More
Mac-Mod Princeton HA-DEA (Diethylamino) 60 Å, 5 µm, 21.2 x 250 mm SFC Column
SDP
Supplier:  Mac-Mod 25021201565
Encompass
Princeton Chromatography Achiral SFC columns are designed and manufactured in the US, well known for innovating novel and robust workhorse phase chemistries, for the analysis of achiral SFC applications. Princeton designs myriad phase chemistries to improve the selectivity and efficiency of SFC analysis. Known for originally innovating the 2-EP phase chemistry, explore what Princeton has to offer. Columns are available from analytical to preparative sizes and are available in a wide range of pore sizes, particle sizes, column ID's and lengths. If you do not see something that you want, please just ask!
Catalog No. 50-001-47112
Add to Cart
Request a Quote