Learn More
Mac-Mod MONOBIO AC rPA 1000 Å, 15 µm, 2.1 x 30 mm Monodisperse HPLC Column
SDP
Supplier:  Mac-Mod MBACRPA440203
Encompass
MONOBIO Affinity Chromatography Columns based on monodisperse particle technology are used for titer analysis and are bonded with a Protein A directly to the surface of the particle
Catalog No. 50-001-52137