Learn More
Mac-Mod MONOBIO HIC Butyl 1000 Å, 5 µm, 4.6 x 100 mm Monodisperse HPLC Column
SDP
Supplier:  Mac-Mod MBHICHBU424610
Encompass
MONOBIO HIC monodisperse HPLC columns are designed to for the determination of hydrophobicity of biomolecules of varying sizes, as well as the analysis of DAR (drug-to-antibody ratio) when analyzing ADC (antibody-drug conjucates)
Catalog No. 50-001-51292