Learn More
Mac-Mod MONOBIO RP-Protein Butyl NP, 5 µm, 4.6 x 100 mm Monodisperse HPLC Column
SDP
Supplier:  Mac-Mod MBRPPRBUN24610
Encompass
MONOBIO RP-Protein monodisperse particle HPLC columns are an excellent choice for the analysis of large globular proteins, and antibodies. These non-porous particle columns provide excellent resoltion and limit carryover for robust separations.
Catalog No. 50-001-50683