Learn More
Mac-Mod MONOBIO RP-Protein Phenyl 1000 Å, 5 µm, 4.6 x 50 mm Monodisperse HPLC Column
SDP
Supplier:  Mac-Mod MBRPPRPH424605
Encompass
MONOBIO RP-Protein monodisperse particle HPLC columns are an excellent choice for the analysis of large globular proteins, and antibodies. These 1000 Angstrom particle columns provide excellent resoltion and maintain excellent silica structure to ensure long column lifetimes and robust separations.
Catalog No. 50-001-51910