Learn More
Mac-Mod MONOBIO SEC DIP 300 Å, 3 µm, 7.8 x 150 mm Monodisperse HPLC Column
SDP
Supplier:  Mac-Mod MBSECDIP217815
Encompass
MONOBIO Size-Exclusion monodisperse particle HPLC columns is a high-pore volume particle while maintaining a very narrow particle size distriubtion, which leads to excellent reproducible results. Also, these columns are known for minor shedding of the bonded phase thus being excellent choices for MALS detections. Choose the next evolution in SEC UHPLC/HPLC column analysis!
Catalog No. 50-001-50939