Learn More
WELCH MATERIALS INC Welch MaterialsTM UltisilTM XB-C18, 3μm, 4.6×150mm
Supplier:  WELCH MATERIALS INC H0020121041
Encompass_Preferred
HPLC Column, brand: Ultisil, bonding phase: C18, dimension: 4.6×150mm, particle size: 3 μm, USP: L1, surface area: 320 m2/g, pore size: 120Å, carbon loading: 17%, pH range: 1.5-10.0, endcapped.
Catalog No. 50-196-3658