Learn More
TrippNT HPLC120 COLUMN CAB W/ HINGED
SDP
Supplier:  TrippNT 51409/NEW
Encompass_Preferred
TrippNT's HPLC Mega 120 Column Cabinet offers secure storage for up to 120 columns with eight removable trays and adjustable dividers. Made from durable white HDPE with clear acrylic lids, this stackable cabinet provides easy inventory visibility. The clear acrylic lids slide forward and backward for convenient access. Designed for customizable storage, it can be wall-mounted using included keyholes and mounting hardware or stacked up to three high for expanded storage. With a hasp lock for security, this cabinet is perfect for organizing HPLC columns in labs. Dimensions: 15 7/8 x 16 1/2 x 17 3/8 inches WHD.
Catalog No. NC3593846
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product