Learn More
Mac-Mod Avantor Alltima HP CN, 100 Å, 3 µm, 4.6 x 50 mm HPLC Column
SDP
Supplier:  Mac-Mod 87825
Encompass
Avantor Alltima HP columns combines a range of different phase chemistries with high purity silica. Avantor Alltima HP columns product family combines the selectivity and performance needed to overcome the most challenging separation needs. Avantor Alltima HP columns provide low column bleed makes these columns ideal for microbore applications. Avantor Alltima HP columns are made of high purity silica that provide excellent column stability with multiple selectivity options.
Catalog No. 50-001-43493