Learn More
Mac-Mod Avantor Apex ODS, 100 Å, 5 µm, 4.6 x 150 mm HPLC Column
SDP
Supplier:  Mac-Mod 4M15310 / 5140900
Encompass
Avantor Apex and Avantor Apex ll phases are recommended for routine analyses. Conventional 100 Å pore size spherical silica particle column. Narrow particle size distribution.
Catalog No. 50-001-43436