Learn More
Mac-Mod Avantor Genesis C8, 120 Å, 4 µm, 3.0 x 30 mm HPLC Column
SDP
Supplier:  Mac-Mod FL3962E
Encompass
Avantor Genesis phases are based on high purity metal-free spherical silica.
Catalog No. 50-001-43322