Learn More
Medchemexpress LLC HPPE | 1325721-55-8 | C21H20F3N5O3S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY153040
Encompass_Preferred
HPPE (compound 236) is an orally active, potential Bach1 inhibitor. Bach1 is a transcription factor that regulates mitochondrial metabolism and reduces glucose utilization. This compound can be used for research in psoriasis, multiple sclerosis, and COPD.
  • Orally active Bach1 inhibitor
  • Modulates mitochondrial metabolism
  • Reduces glucose utilization
  • Supports research in psoriasis, multiple sclerosis, and COPD
  • Induces Bach1 derepression
  • Stabilizes Nrf2 activity
  • Activates NRF2 driven transcription
  • Ameliorates dopaminergic neurodegeneration in animal models
Catalog No. 50-003-74340