Learn More
MSC PIP Cut-Resistant Gloves, Nitrile Coating, 13 Gauge, 2X-Large, 10/Pk.
Supplier: MSC 00991471
These cut-resistant gloves provide advanced mechanical protection with a 13-gauge engineered yarn liner, designed for applications requiring high dexterity and comfort. Featuring a smooth nitrile coating, these gloves offer a reliable dry grip and effective protection against abrasions, punctures, and tears in industrial environments.
- Engineered yarn construction ensures one of the lightest cut-protective solutions for reduced hand fatigue
- Skin-friendly materials enhance comfort and minimize allergic reactions
- Reinforced thumb crotch improves durability in high-wear areas
- Touchscreen compatible design for operational efficiency
- Extended wrist cuff for increased coverage
- OEKO-TEX 100 certified and silicone-free
- ANSI/ISEA cut resistance level A4 and EN 388 rating of 4X44D