Learn More
MSC PIP Perfect Fit 3.0 Cut Resistant Gloves, Cut A4, 18 ga, Small, 10/Pk.
Supplier: MSC 05771431
These cut, puncture and abrasion-resistant gloves provide advanced hand protection for diverse industrial applications. Engineered with a lightweight 18 gauge seamless liner constructed from HPPE, nylon, carbon, and elastane, these gloves offer superior dexterity and comfort. The palm and fingertips are coated with black polyurethane for grip performance and breathability. These gloves are antistatic, touchscreen capable, and certified for food handling.
- ANSI/ISEA cut level A4 protection
- ANSI/ISEA abrasion resistance level 3
- ANSI/ISEA puncture resistance level 3
- Lightweight 18 gauge seamless liner
- Polyurethane palm and fingertip coating
- Antistatic and touchscreen capable
- Silicone-free and glass fiber-free construction
- Certified for food contact under FDA and EC regulations
- Low DMF content less than 100 ppm
- Washable for up to 3 cycles