Learn More
Medchemexpress LLC HPPH (Photochlor) | 149402-51-7 | MFCD11040935 | 99.7% | C39H48N4O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1372210MG
Encompass_Preferred
HPPH (Photochlor) is a second-generation, chlorin-based photosensitizer used in photodynamic therapy research. It selectively accumulates in tumor tissue and, when activated by red light (≈655 nm), generates reactive oxygen species that induce localized cytotoxicity. The compound is supplied as a solid for laboratory use and is intended for research purposes only.
  • Chlorin-based photosensitizer suitable for photodynamic therapy research.
  • Activates with red light around 655 nm to produce reactive oxygen species.
  • Selective accumulation in tumor tissue enabling targeted cytotoxic effects.
  • Available as a solid for laboratory experimental use.
  • Documented chemical identity and physical properties for reproducible experiments.
Catalog No. 50-002-59380