Learn More
Lonza Walkersville HprAD - Human Subcutaneous Preadipocyte Cells
Supplier:  Lonza Walkersville PT5001
Encompass
Poietics Normal Human Preadipocytes (HprAD) are isolated from subcutaneous or visceral fat from normal (non-diabetic) donors. Preadipocytes from diabetic donors are available as a separate product.Pertinent additional donor information, including Body Mass Index (BMI) may be available by contacting Scientific Support.Cryopreserved Poietics Normal Human Preadipocytes (HprAD) are guaranteed to contain ≥1 million or ≥4 million (subcutaneous only) cells per ampoule and test positive for adipogenic differentiation. Benefits:Test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsA Certificate ofAnalysis is provided for each cell lot purchased Content:1 x Cryopreserved ampule of Subcutaneous Preadipocytes containing ≥ 4 million cells
Catalog No. NC2161022