Learn More
Microsolv Technology Corporation Hps C18 4.6X150Mm
Supplier:  Microsolv Technology Corporation 7501815P
Encompass_Preferred
HPLC Column, HPS C18, 5um, 4.6mm ID x 150mm Length, 120A. Cogent Brand. 1/EA.
Catalog No. 50-014-51395