Learn More
Medchemexpress LLC HPSE Rat His-Myc 50ug
SDP
Supplier:  Medchemexpress LLC HYP7223550UG
Encompass_Preferred
HPSE proteins cleave HSPG into heparan sulfate side chains and decorin involved in ECM degradation and remodeling HPSE promotes cell migration associated with metastasis wound healing and inflammation and acts as a procoagulant HPSE also induces AKT1/PKB phosphorylation enhancing angiogenesis hair follicle differentiation and homeostasis HPSE Protein Rat (His-Myc) is the recombinant rat-derived HPSE protein expressed by E coli with N-10 His C-Myc labeled tag
Catalog No. 50-001-81350