Learn More
BIOHIPPO hPUMA-sh2-GFP/Luc
SDP
Supplier:  BIOHIPPO BHV194001545
Encompass
hPUMA-sh2-GFP/Luc shRNA lentiviral vector expressing GFP/Luc with selection for studies in Human Each vial provides a nominal titer of 2x10E6 transducing units TU Validated human PUMA shRNA lentivirus High-titer shRNA lentiviral particles specific for human PUMA also known as BCL2 binding component 3 (BBC3) JFY1 or JFY-1 The shRNA has been validated to meet or exceed 70% PUMA knockdown efficiency using a specific fluorescence-based method that is more rapid and reliable than qPCR The shRNA lentivirus is ultra-purified and concentrated to high-titer by PEG precipitation and sucrose gradient centrifugation and ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs
Catalog No. 50-003-31030