Learn More
Enzo Life Sciences PATHO-GENE HPV screening probe (6ml)
SDP
Supplier:  Enzo Life Sciences ENZ328846000
Encompass_Preferred
The PATHO-GENE HPV screening probe is a mixture of biotin-labeled HPV 6, HPV 11, HPV16, HPV 18, HPV 31, HPV 33, and HPV 51-specific probes in buffered formamide and hybridization enhancers. Long Term Storage: +4°C.
Catalog No. 50-201-2493