Learn More
Creative Biomart Recombinant HPV-16 E6 Antigen, His-tagged
Supplier:  Creative Biomart HPV348
Encompass_Preferred
Recombinant Papillomavirus 16 E6 antigen produced in E. coli contains 159 amino covering the full length of HPV-16 having its C-terminus fused with a 6 His tag and a total molecular weight of 21 kD
Catalog No. 50-208-4726