Learn More
New England Biolabs, Inc. Hpy188III – 2500 units
SDP
Supplier:  New England Biolabs, Inc. R0622L
Encompass
A restriction endonuclease that recognizes the sequence TC^NN_GA.
Catalog No. 50-812-361